Anti-Stromal Cell Derived Factor 1 beta Goat Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBIS7975-62FEA 738 EUR
USBIS7975-62F
Anti-Stromal Cell Derived Factor 1 beta Goat Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 0.15-0. 3?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Stromal Cell Derived Factor 1 beta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR