Anti-T Cell Adaptor Protein Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIT2015EA 480 EUR
USBIT2015
Anti-T Cell Adaptor Protein Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: T Cell Adaptor Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR