Anti-Stromal Cell Derived Factor 1 beta Goat Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBIS7975-62C-50EA 470 EUR
USBIS7975-62C-50 USBIS7975-62C-25
Anti-Stromal Cell Derived Factor 1 beta Goat Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: Stromal Cell Derived Factor 1 beta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR