Anti-Proliferating Cell Nuclear Antigen Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI141981EA 459 EUR
USBI141981
Anti-Proliferating Cell Nuclear Antigen Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Proliferating Cell Nuclear Antigen
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR