Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: 66.4.C2, PN-15]

Supplier: United States Biological
USBI138592EA 608 EUR
USBI138592
Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: 66.4.C2, PN-15]
Antibodies
Recommended dilution: ;Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Renal Cell Carcinoma
Clonality: Monoclonal
Clone: 66.4.C2, PN-15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR