Anti-PTBP3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP17726AEA 348 EUR
ABGEAP17726A
Anti-PTBP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR