Anti-CEACAM7 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP17149CEA 348 EUR
ABGEAP17149C
Anti-CEACAM7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CEACAM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR