Anti-CIDEC Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP18249CEA 348 EUR
ABGEAP18249C
Anti-CIDEC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR