Anti-SUMO-1 Sheep Polyclonal Antibody

Supplier: Biosensis
BSENS-064-100EA 312 EUR
BSENS-064-100
Anti-SUMO-1 Sheep Polyclonal Antibody
Antibodies
SUMO-1 binds to a wide range of target proteins as part of a post-translational modification system. Unlike ubiquitin, it does not seem to target protein for degradation, but is involved in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis, apoptosis, protein stability and signal transduction. SUBUNIT: Covalently attached to a number of proteins such as PML, RANGAP1, HIPK2, SP100, p53, p73alpha, MDM2, JUN and DNMT3B. Also interacts with HIF1A, HIPK2, HIPK3, CHD3, PIAS1, EXOSC9, TDG, RAD51 and RAD52. SUBCELLULAR LOCATION: Nucleus; nuclear membrane. Nucleus; nucleoplasm; nuclear speckle. Cytoplasm. SIMILARITY: Belongs to the ubiquitin family. SMT3 subfamily. SIMILARITY: Contains 1 ubiquitin-like domain. PTM: Cleavage of the last four amino acids of the carboxy-terminus of the precursor form by SENP1 or SENP2 is necessary for function. Several pseudogenes have been reported as well as a number of alternatively spliced isoforms.

This antiserum recognises human SUMO-1 and not ubiquitin.
Application Information:
IHC, WB. A dilution of 1:2000 to 1:4000 is recommended for immunohistochemistry and 1:4000 to 1:8000 for western blot. Cell lysate from Hela and NIH-3T3 cell lysates may be used as a positive control, and for IHC, lung carcinoma may be used. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SUMO-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human SUMO-1 peptide (aa: 6-21)
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR