NT3 is a member of the neurotrophin family, that controls survival and differentiation of visceral and proprioceptive sensory neurons. NT3 is closely related to both NGF and BDNF. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NT3-deficient mice generated by gene targeting display sevvere movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family
The antibody reacts with human and mouse NT-3 proteins and likely with other species based upon homology. The antibody has not been tested in IHC or western blot
Application Information:
ELISA at suggested dilution of 1:500-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Neurotrophin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human NT3 peptides, amino acids 90-104 and 199-214.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse