Clifton® block heaters provide a safe, dry and consistent method of heating accessory aluminium blocks with excellent thermal conductivity for heating tubes. Features a bold LED display of measured temperatures in real-time, increments of ±0,1 °C, using precision PID temperature control. Setting for block temperature only or combination of temperature and time.
- Easy to use controls, touch keypads and indicators for heating, over-temperature alarm and timer
- Settings are retained even after turning the unit off
- Thermal over temperature protection, with auto-setting over-temperature alarm
Delivery information: Supplied with a block extraction tool.