dNTP mix, peqGOLD

Supplier: VWR Life Science
PEQL20-3010EA 20.1 EUR
PEQL20-3010
dNTP mix, peqGOLD
dNTP mix contains a ready-to-use mixture of dATP, dCTP, dGTP and dTTP. Each synthetically manufactured deoxynucleotide has a concentration of 10 mM. Available in 4 quantities 2 µmol, 10 µmol, 50 µmol and 250 µmol of each individual nucleotide, ie. 8 µmol, 40 µmol, 200 µmol and 1000 µmol mixed dNTPs.

  • Purity, measured by HPLC, >99%
  • Super-high stability with >90% of peqGOLD dNTPs stable after 6 weeks storage at room temperature
  • pH 7,5 to prevent hydrolysis of phosphate groups during long-term storage
  • Manufacturing and quality control under ISO 9001 and ISO 14001
  • Tested free from 5'-exonucleases, 3'-exonucleases, non specific endonucleases, phosphatase contaminants, and RNases
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR