Potassium thiocyanate ≥98.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™

7168-04
Potassium thiocyanate ≥98.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Potassium thiocyanate
Formula: KSCN
MW: 97,18 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1,61 g/cm³ (20 °C)
MDL Number: MFCD00011413
CAS Number: 333-20-0
Merck Index: 12,07866

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KSCN) (by Ag titrn) ≥98.5 %
Solubility (10 in 50) Passes Test
ACS - Ammonium (NH₄) ≤0.003 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble in H₂O ≤0.005 %
ACS - Iodine Consuming Substances (meq/g) ≤0.004
ACS - Iron (Fe) ≤0.0002 %
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.3 - 8.7
Appearance (colorless or white crystals)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR