Inlet liners, Supelco®
Supplier: Merck
2878301-UEA 41.1 EUR
2878301-U 2878305-U
Inlet liners, Supelco®
Chromatography Supplies Chromatography Inlet Liners
These are deactivated (silanised) glass inlet liners for capillary injection ports.
  • Fits Shimadzu