Gold for bright brushing

Supplier: Thermo Scientific
Danger

12943.04EA 87.5 EUR
12943.04
Gold for bright brushing
Gold

Bright brushing gold.


Produces a bright continuous gold film on glass and ceramics when properly fired. Firing instructions included.

Formula: Au
MW: 196,97 g/mol
Boiling Pt: 156 °C
Flash Pt: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR