Dibutyl phosphate ≥96%
Supplier: Thermo Scientific
|
Danger
|
43371.AKEA
79.1
EUR
43371.AK
Dibutyl phosphate ≥96%
Dibutyl phosphate
Formula:
C₈H₁₉O₄P MW: 210,21 g/mol Boiling Pt: 250 °C Melting Pt: –13 °C Density: 1,058 Flash Pt: 157 °C (315 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015245 CAS Number: 107-66-4 EINECS: 203-509-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...