Palladium 1% on alumina, spheres 1-2 mm, reduced

44820.36EA 1960 EUR
44820.36
Palladium 1% on alumina, spheres 1-2 mm, reduced
Palladium
Formula: Pd
MW: 106,420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR