Thyrotropin-releasing hormone

Supplier: Thermo Fisher Scientific
J62120.MFEA 96.1 EUR
J62120.MF
Thyrotropin-releasing hormone
Proteins and Peptides
CAS No.: 24305-27-9
Molecular Formula: C16H22N6O4
Formula Weight: 362,38
Storage Temperature: −30 to −10 °C
Physical Form: Powder
Appearance: White to pale yellow
Beilstein Reference: 770238
MDL No.: MFCD00038640

Applications: Two-chain glycoprotein hormone. Activates adenylate cyclase in the thyroid gland, stimulating iodine uptake, thyroxine synthesis and release
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR