Streptavidin (from Streptomyces avidinii)

Supplier: Thermo Fisher Scientific
J62428.MAEA 350 EUR
J62428.MA
Streptavidin (from Streptomyces avidinii)
Proteins and Peptides
Purity: 95%
CAS No.: 9013-20-1
Storage Temperature: −30 to −10 °C
Physical Form: Lyophilised powder, 13,5 kDa
Appearance: White
MDL No.: MFCD00082035

Applications: A protein with high affinity for biotin
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR