Tripeptidyl peptidase Inhibitor

Supplier: Thermo Fisher Scientific
J64760.MAEA 217 EUR
J64760.MA
Tripeptidyl peptidase Inhibitor
Proteins and Peptides
Applications: A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II

CAS No.: 184901-82-4
Molecular Formula: C16H22ClN3O3
Formula Weight: 453,90
Storage Temperature: −30 to −10 °C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD01310850
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR