Boc-D-glutamine ≥98%

Supplier: Thermo Scientific

L08536.03EA 60 EUR
L08536.03
Boc-D-glutamine ≥98%
Boc-D-glutamine
Formula: C₁₀H₁₈N₂O₅
MW: 246,263 g/mol
Melting Pt: 117…119 °C
CAS Number: 61348-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR