Process supplement, HyClone Cell Boost™ 2
Supplier: HyClone products (Cytiva)
SH30596.01EA 196.55 EUR
SH30596.01
Process supplement, HyClone Cell Boost™ 2
Cell Culture Supplements and Additives
HyClone™ Cell Boost™ 2 Supplement is designed to provide nutrients such as amino acids and glucose to support fed-batch cell culture.
  • Chemically defined, protein-free and free of animal-derived components (ADCF)
  • Tested on a variety of cell lines, including CHO and PER.C6
  • No L-glutamine or poloxamer 188
  • Ideal for supplementing nutrients for bioprocessing and media customisation applications
  • Manufactured according to cGMP guidelines