Process supplement, HyClone Cell Boost™ 1
Supplier: HyClone products (Cytiva)
HYCLSH30584.02EA 225.72 EUR
HYCLSH30584.02
Process supplement, HyClone Cell Boost™ 1
Cell Culture Supplements and Additives
HyClone™ Cell Boost™ 1 Supplement is designed to provide nutrients such as amino acids, vitamins and glucose as part of fed-batch cell culture strategies.
  • Chemically defined, protein-free and free of animal-derived components (ADCF)
  • Tested on a variety of cell lines, including Chinese hamster ovary (CHO) and HEK 293 cells
  • No L-glutamine or poloxamer 188
  • Ideal for supplementing nutrients for bioprocessing and media customisation applications
  • Manufactured according to cGMP guidelines