Cell culture media for insect cell culture, Hyclone™
Supplier: HyClone products (Cytiva)
HyClone™
HYCLSH30282.01EA 0 EUR
HYCLSH30282.01
Cell culture media for insect cell culture, Hyclone™
Cell Culture Media
Classical media formulations for insect cell culture, used in basic cell biology research and for the efficient expression of proteins using Baculovirus expression systems.
  • Liquid media produced with WFI quality water and are 0,1 μm sterile filtered
  • Do not contain phenol red
  • Manufactured using ISO 9001:2000 certified processes
IPL-41: IPL-41 insect medium was originally designed for the growth of Spodeptera frugiperda (Sf9) cells. IPL-41 media contains essential ingredients for insect cell culture, including malic acid, fumaric acid, and alpha-ketoglutaric acid. IPL-41 is a basal medium frequently used for serum-free media development and optimisation.