Alkylation reagents
Supplier: Thermo Scientific
HYPETS-49350EA 503 EUR
HYPETS-49350
Alkylation reagents
Chromatography Sorbents GC Sorbents
Methylate Reagent (DMFDMA) reactions are typically complete upon dissolution (except for long-chain solid acids) and quantitative yields are obtained when reagent and sample are injected without prior mixing.
  • Ready-to-use reagent
  • For easy and effective preparation of methyl esters from fatty acids and amino acids
  • No water washing, extraction or concentration of derivatives required
Packaging: Packaged in hypo-vial sample storage vial.