Anti-TRAIL Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ALEXADI-AAP-470-EEA 517 EUR
ALEXADI-AAP-470-E
Anti-TRAIL Rabbit Polyclonal Antibody
Antibodies
TRAIL is a type II membrane protein and expressed in a variety of human tissues.

  • Dilution range: Immunohistochemistry (20 µg/ml), Western blot (1 µg/ml, ECL)
  • Detects bands of ~32 kDa and ~24 kDa (membrane and soluble forms) by Western blot

Two death domain containing receptors, DR4 and DR5, have been identified as the receptors for TRAIL. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-κB activation in many tissues and cells.

Type: Primary
Antigen: Tumor Necrosis Factor (Ligand) Superfamily, Member 10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR