Anti-Cancer Cell Development Antibody

Supplier: Novus Biologicals
NOVUNBP1-78940EA 299 EUR
NOVUNBP1-78940
Anti-Cancer Cell Development Antibody
Antibodies
Cancer Cell Development Antibody Pack. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type:
Antigen: Cancer Cell Development
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: 0
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR