Anti-Pluripotent Stem Cell Transcription Factor Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNBP1-42823EA 1030 EUR
NOVUNBP1-42823
Anti-Pluripotent Stem Cell Transcription Factor Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Pluripotent Stem Cell Transcription Factor Antibody Pack. Tested Applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactiv

Type:
Antigen: Pluripotent Stem Cell Transcription Factor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR