Cell lysis buffer, pH 7,5
Supplier: Enzo Life Sciences
ALEXADI-80-1134EA 234 EUR
ALEXADI-80-1134
Cell lysis buffer, pH 7,5
Buffers Extraction Buffers
The liquid cell lysis buffer composes of 100 mM TRIS, pH 7,5, containing 2 mM EDTA, 2 mM EGTA, 5 mM beta-glycerophosphate, 20 mM sodium pyrophosphate, 0,1% Triton X-100, 0,1% Tween 20 and 0,1% Hydorol M.
Storage temperature: 4 °C