Cell lysis buffer, pH 7,4
Supplier: Enzo Life Sciences
ALEXADI-80-1631EA 264 EUR
ALEXADI-80-1631
Cell lysis buffer, pH 7,4
Buffers Extraction Buffers
The liquid cell lysis buffer composes of 20 mM TRIS, pH 7,4, containing 500 mM sodium chloride, 10 mM EDTA, 1% Triton X-100, 20 mM sodium fluoride, 20 mM beta-glycerophosphate.
Storage temperature: 4 °C