Orthoboric acid ACS

Supplier: MP Biomedicals
Danger

ICNA0219141101EA 54 EUR
ICNA0219141101 ICNA0219141105 ICNA02191411.1 ICNA02191411.5
Orthoboric acid ACS
Orthoboric acid

Boric acid is used for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems; in nickeling baths; cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel. It is also used as an insecticide for cockroaches and black carpet beetles. It is used In the research labs , astringent, antiseptic, antibacterial and antifungal agent. It is used in buffers and for calibration of polyacrylamide gel columns for the separation of oligonucleotides by capillary electrophoresis.

Boric acid has a single boron; its salt forms complex equilibria in solution, with the ion B4O7-2 being the predominant species.

Formula: B(OH)₃
MW: 61,83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
https://export.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR