Anti-TNFRSF9 Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI36-216EA 454 EUR
PRSI36-216
Anti-TNFRSF9 Rabbit Polyclonal Antibody
Antibodies
CD137 is a member of the tumor necrosis factor (TNF) receptor family. CD137 is expressed by activated T cells, dendritic cells, NK cells, granulocytes and cells of blood vessel walls at sites of inflammation. CD137 has costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. CD137 was shown to enhance immune activity to eliminate tumors in mice.

Flow Cytometry: (1:500-1:1,000). Western Blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is TBST with 0.05% Tween 20 with 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR