Mouse recombinant Host cell factor 2 (from E. coli)
Supplier: ProSci
PRSI92-303EA 487 EUR
PRSI92-303
Mouse recombinant Host cell factor 2 (from E. coli)
Proteins and Peptides
Host cell factor 2(HCFC2) is a cytoplasmic protein. It contains 2 fibronectin type-III domains.HCFC2 binds KMT2A/MLL1, as component of the MLL1/MLL complex.Hcfc2 negative regulation of transcription from RNA polymerase II promoter.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.