Human recombinant SCF (from CHO cells)
Supplier: ProSci
PRSI90-538EA 644 EUR
PRSI90-538
Human recombinant SCF (from CHO cells)
Proteins and Peptides
Stem cell factor (SCF), also known as cKit ligand (KL), mast cell growth factor (MGF) and steel factor (SLF), is a widely expressed 28-40 kDa type I transmembrane glycoprotein. It promotes the survival, differentiation and mobilisaion of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell and melanocyte progenitors. SCF is a primary growth and activation factor for mast cells and eosinophils. Noncovalent dimers of transmembrane or soluble SCF interact with the receptor tyrosine kinase SCF R/cKit to trigger receptor dimerisaion and signalling. SCF assists in the recovery of cardiac function following myocardial infarction by increasing the number of cardiomyocytes and vascular channels.
  • Fusion-Tag: Fc Tag
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.