Anti-CCR3 Goat Polyclonal Antibody

Supplier: Enzo Life Sciences
ALEXALX210641C200EA 574 EUR
ALEXALX210641C200
Anti-CCR3 Goat Polyclonal Antibody
Antibodies
Host: Goat

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes mouse CCR3.

Type: Primary
Antigen: CCR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR