Anti-NOD1 Rabbit Polyclonal Antibody

ALEXALX210918C050EA 324 EUR
ALEXALX210918C050
Anti-NOD1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human Nod1.

Type: Primary
Antigen: NOD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR