Anti-BAFFR Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
ALEXALX803332C100EA 524 EUR
ALEXALX803332C100
Anti-BAFFR Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2b

Recognizes mouse BAFF-R.

Type: Primary
Antigen: BAFFR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR