Anti-ISG15 Rabbit Polyclonal Antibody

Supplier: Axxora
ALEXBMLPW95750100EA 416 EUR
ALEXBMLPW95750100 ALEXBMLPW95750025
Anti-ISG15 Rabbit Polyclonal Antibody
Antibodies
ISG15, also called ubiquitin cross-reactive protein (UCRP), is an interferon stimulated ubiquitin-like protein. Found only in higher eukaryotic organisms, it may play important roles in viral or bacterial infection where protein ISGylation is highly induced.

  • For Western Blot

Recognizes human ISG15/UCRP.

Type: Primary
Antigen: ISG15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR