Anti-Surfactant Protein D Mouse Monoclonal Antibody

Supplier: Axxora
ALEXBPDHYB2450202EA 636 EUR
ALEXBPDHYB2450202
Anti-Surfactant Protein D Mouse Monoclonal Antibody
Antibodies
Surfactant protein A (SP-A) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins, members of which possess an overall structure consisting of multiple globular head regions linked by triple-helical, collagen-like, strands. This group also includes SP-D, and the serum proteins mannan-binding protein, conglutinin, and collectin-43, all of which bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A enhance oxygen radical production by alveolar macrophages. The serum concentration is 45 ng/mL in healthy individuals.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human surfactant protein D.

Type: Primary
Antigen: SURFACTANT PROTEIN D
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR