Anti-CK1 Mouse Monoclonal Antibody [clone: 34BE12]

Supplier: Axxora
ALEXENZ-30903EA 341 EUR
ALEXENZ-30903
Anti-CK1 Mouse Monoclonal Antibody [clone: 34BE12]
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes human high molecular weight cytokeratins 1, 5, 10 and 14.

Type: Primary
Antigen: CK1
Clonality: Monoclonal
Clone: 34ΒE12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR