Anti-KLK8 Mouse Monoclonal Antibody [clone: B12]

Supplier: Axxora
ALEXADI90510050100EA 639 EUR
ALEXADI90510050100
Anti-KLK8 Mouse Monoclonal Antibody [clone: B12]
Antibodies
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK8 is one of the fifteen kallikrein subfamily member. The KLK8 enzyme exhibits distinct patterns of expression that suggest roles in brain plasticity and ovarian cancer.

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR