Anti-BAG4 Rabbit Polyclonal Antibody

Supplier: Axxora
ALEXADI905243100EA 591 EUR
ALEXADI905243100
Anti-BAG4 Rabbit Polyclonal Antibody
Antibodies
The TNF-R1 interacting protein SODD (silencer of death domains) associates with the death domain of TNF-R1 and prevents constitutive activation of TNF-R1 signaling.

Type: Primary
Antigen: BAG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR