Anti-LIMK2 Rat Monoclonal Antibody [clone: 10E5-19]

Supplier: Axxora
ALEXALX803344C100EA 275 EUR
ALEXALX803344C100
Anti-LIMK2 Rat Monoclonal Antibody [clone: 10E5-19]
Antibodies
The LIM kinases 1 and 2 (LIMK1 and LIMK2) are activated downstream of the Rho-ROCK, Rac-PAK, and Cdc42-MRCK pathways, thereby acting as network nodes in the signaling pathways that regulate the actin cytoskeleton. LIMK is required for matrix remodeling activities for path generation by leading cells in collective invasion.

Type: Primary
Antigen: LIMK2
Clonality: Monoclonal
Clone: 10E5-19
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR