Anti-IgG Rabbit Polyclonal Antibody

Supplier: Axxora
ALEXENZABS2641000EA 493 EUR
ALEXENZABS2641000
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Recommended Applications: WB

Species reactivity: Human

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR