Cell migration BioGel HT assay, TC
Supplier: Enzo Life Sciences
ALEXENZKIT1110001EA 447 EUR
ALEXENZKIT1110001 ALEXENZKIT1110005
Cell migration BioGel HT assay, TC
Assays Cellular Assays
Reproducibly monitor real-time cell migration, with a high-throughput platform.
2D high-throughput assay for investigating cell migration of adherent cell lines.
The Cell migration BioGel HT assay is a reproducible, sensitive, and flexible assay that can be used to monitor cell migration. Formatted in a 384-well plate, the assay uses a non-toxic biocompatible gel (BioGel) to form a cell-free zone. After seeding cells into the 384-well plate, the BioGel self-dissolves permitting cells to migrate into the well centers.The Cell migration BioGel HT assay enables the use of automated liquid handling equipment for cell seeding and allows for unlimited access to wells from cell seeding through data readout. The Cell migration BioGel HT assay is designed to be used with any stain or labeling technique. Researchers can capture and quantify real-time and endpoint cell migration data using inverted microscopes, High Content Screening (HCS) and High Content Imaging (HCI) instruments.The Cell migration BioGel HT assay system has been designed for use with adherent cell cultures. This assay has been successfully used with HT-1080 and MDA-MB-231 cell lines, and primary Human Umbilical Vein Endothelial Cells (HUVECs).
Ordering information: Contains -0001 size:1 x 384 well plate - Cell migration BioGel HT assay plate, TC (Tissue Culture treated)-0005 size:5 x 384 well plates - Cell migration BioGel HT assay plate, TC (Tissue Culture treated).