Cell migration BioGel HT assay, Collagen-I coated
Supplier: Enzo Life Sciences
ALEXENZKIT1120001EA 479 EUR
ALEXENZKIT1120001 ALEXENZKIT1120005
Cell migration BioGel HT assay, Collagen-I coated
Assays Cellular Assays
Reproducibly monitor real-time cell migration, with a high-throughput platform.
2-D High-Throughput assay for investigating cell migration of adherent cell lines
The Cell migration BioGel HT assay, Collagen-I coated is a reproducible, sensitive, and flexible assay that can be used to monitor cell migration. Formatted in a 384-well plate, the assay uses a non-toxic biocompatible gel (BioGel) to form a cell-free zone. After seeding cells into the 384-well plate, the BioGel self-dissolves permitting cells to migrate into the well centers.The Cell migration BioGel HT assay, Collagen-I enables the use of automated liquid handling equipment for cell seeding and allows for unlimited access to wells from cell seeding through data readout. The Cell migration BioGel HT assay, Collagen-I is designed to be used with any stain or labeling technique. Researchers can capture and quantify real-time and endpoint cell migration data using inverted microscopes, High Content Screening (HCS) and High Content Imaging (HCI) instruments.The Cell migration BioGel HT assay, Collagen-I system has been designed for use with adherent cell cultures. This assay has been successfully used with HT-1080 and primary Human Umbilical Vein Endothelial Cells (HUVECs).
Ordering information: -0001 size contains:1 x 384 well plate - Cell migration BioGel HT assay plate, Collagen-I coated. -0005 size contains: 5 x 384 well plates - Cell migration BioGel HT assay plate, Collagen-I coated.