DiSH™ kit
Supplier: Enzo Life Sciences
ALEXENZ-71001-0001EA 1640 EUR
ALEXENZ-71001-0001
DiSH™ kit
Assays Immunological Assays
Reduce labor time and costs using a novel technology for rapid hybridoma production
  • Unique technology for more rapid hybridoma production
  • ecreased time and labor costs
  • Improved selection of specific, stable hybridomas
  • Minimal manipulation of cloned cell lines relative to approaches requiring Limiting Dilution Subcloning
Uses:
Hybridoma generation and Cloning
Cell line development
Recommended for generation, selection and cloning of mouse monoclonal hybridomas.
A genetically-modified myeloma partner which creates hybridomas that both secrete antigen-specific antibody and also express antibody on the cell surface of the hybridoma.
The DiSH Kit contains Abeome's patented SP2ab myeloma fusion partner PLUS culture, fusion, selection and recovery media for creation and isolation of hybridomas by fluorescent or magnetic separation.
Ordering information: Contains 1 vial of Sp2ab myeloma partner cells, 1.5 ml PEG, 200 ml PCM (Prefusion Medium), (2X) 200 ml WCM (Wash Medium), 200 ml FCM (Fusion Medium), 90 mL AbeoClone™ Medium, (2X) 200 ml RCM (Recovery Medium), 200 ml ECM (Expansion Medium).