Indirubin-5-sulphonic acid sodium salt
Supplier: Enzo Life Sciences
Synonyms:
Indirubin-5-sulphonate Na
, E226
ALEXALX270296M005EA
520
EUR
ALEXALX270296M005
ALEXALX270296M001
Indirubin-5-sulphonic acid sodium salt
Indirubin-5-sulphonic acid sodium salt
Inhibitor of cyclin-dependent kinases (CDKs), with selectivity for CDK1/cyclin B (IC50=55 nm), CDK2/cyclin A (IC50=35 nm), CDK2/cyclin E (IC50=150 nm), CDK4/cyclin D1 (IC50=300 nm), CDK5/p35 (IC50=65 nm) and GSK-3β (IC50=280 nm).