3-O-Methyl-N-acetyl-D-glucosamine ≥98% (by 1H-NMR)

ALEXALX270343M005EA 387 EUR
ALEXALX270343M005
3-O-Methyl-N-acetyl-D-glucosamine ≥98% (by 1H-NMR)
3-O-Methyl-N-acetyl-D-glucosamine

N-Acetylglucosamine kinase inhibitor


Inhibitor of N-acetylglucosamine kinase (Ki=17µM) and N-acetylmannosamine kinase (Ki=80µM). It has been found that 3-OMe-GlcNAc potently inhibits glucose phosphorylation by N-acetylglucosamine kinase whereas glucokinase is not at all affected by this hexosamine.

Formula: C₉H₁₇NO₆
MW: 235,2 g/mol
CAS Number: 94825-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR