Indirubin ≥98% (by 1H-NMR, HPLC)

Supplier: Enzo Life Sciences

ALEXALX270361M005EA 443 EUR
ALEXALX270361M005 ALEXALX270361-M001
Indirubin ≥98% (by 1H-NMR, HPLC)
Indirubin

Inhibitor of CDK and GSK


Inhibitor of cyclin-dependent kinases (CDK1/cyclin B (IC50=10µM), CDK2/cyclin A (IC50=2.2µM), CDK2/cyclin E (IC50=7.5µM), CDK4/cyclin D1 (IC50=12µM), CDK5/p35 (IC50=5.5µM)) and of GSK-3beta (IC50=600nM). Inhibits inflammatory reaction in delayed-type hypersensitivity in mice. Ligand of human aryl hydrocarbon receptor. Exhibits anti-cancer activity.


Synthetic.

Formula: C₁₆H₁₀N₂O₂
MDL Number: MFCD00221745
CAS Number: 479-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR